Learn More
C3orf10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16821695 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16821695 150 μL 150µL
16831695 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16821695 Supplier Proteintech Supplier No. 221571AP150UL
Rabbit Polyclonal Antibody
C3orf10 is involved in regulation of actin and microtubule organization. It is a part of a WAVE complex that activates the Arp2/3 complex.
Specifications
Antigen C3orf10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene BRK1
Gene Accession No. Q8WUW1, Q91VR8
Gene Alias BRK1, C3orf10, hHBrk1, HSPC300, MDS027, Probable protein BRICK1
Gene Symbols BRK1
Host Species Rabbit
Immunogen C3orf10 Fusion Protein Ag17793
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 101314, 55845, 679934
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG