Learn More
C1orf116 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14888-1-AP-20UL
 View more versions of this product
Product Code. 16803784
  • 145.00€ / 20µL
Description
Two isoforms of SARG are produced by alternative splicing encoding 64 kDa and 38 kDa proteins, and both isoforms can be recognized by this antibody (Product No. 14888-1-AP).
Specifically androgen-regulated gene protein (SARG), encoded by C1orf116, is a 601-aa protein considered putative androgen-specific receptor. SARG mRNA is highly expressed in prostate and can be up-regulated by androgens, but not by glucocorticoids. Little is known about the function of SARG to date.
Specifications
Specifications
C1orf116
Polyclonal
Unconjugated
SARG
C1orf116, SARG
Rabbit
Antigen Affinity Chromatography
RUO
212439, 79098
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BW04
AA986860, SARG
C1orf116 Fusion Protein Ag6675
20 μL
Primary
Human, Mouse
Antibody