missing translation for 'onlineSavingsMsg'
Learn More

C18orf8 Monoclonal Antibody (OTI4E4), TrueMAB™, OriGene

Product Code. 17501912
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17501912 100 μL 100µL
17521912 30 μL 30µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17501912 Supplier Origene Technologies Supplier No. TA504561

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Colon cancer-associated protein Mic1.

Specifications

Antigen C18orf8
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI4E4
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene C18orf8
Gene Accession No. Q96DM3
Gene Alias C18orf8, HsT2591, MIC1, Mic-1, WDR98
Gene Symbols C18orf8
Host Species Mouse
Immunogen Full length human recombit protein of human C18orf8 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29919
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.