missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C13orf37 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24865-1-AP-20UL
This item is not returnable.
View return policy
Description
Required for gamma-tubulin complex recruitment to the centrosome.Specifications
| C13orf37 | |
| Polyclonal | |
| Unconjugated | |
| MZT1 | |
| C13orf37, MOZART1, MZT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 440145 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q08AG7 | |
| MZT1 | |
| C13orf37 Fusion Protein Ag21699 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction