missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C11orf79 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19906-1-AP-150UL
This item is not returnable.
View return policy
Description
19906-1-AP is a rabbit polyclonal antibody raised against the full-length of human SDHAF2.
SDHAF2, also named asC11orf79, SDHAF2 and SDH5, is a mitochondrial protein needed for the flavination of a succinate dehydrogenase complex subunit required for activity of the complex. SDHAF2 has been shown to be mutated in patients with paragangliomas, also known as familial non-chromaffin paragangliomas type 2.Specifications
| C11orf79 | |
| Polyclonal | |
| Unconjugated | |
| SDHAF2 | |
| C11orf79, hSDH5, PGL2, SDH assembly factor 2, SDH5, SDHAF2, SDHAF2, PGL2, SDH5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361726, 54949, 66072 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5RJQ7, Q8C6I2, Q9NX18 | |
| Sdhaf2 | |
| C11orf79 Fusion Protein Ag13893 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction