missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C. tetani TeNT Light Chain Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6535
This item is not returnable.
View return policy
Description
TeNT Light Chain/Tetanus Toxin Monoclonal specifically detects TeNT Light Chain/Tetanus Toxin in C. tetani samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| TeNT Light Chain/Tetanus Toxin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL | |
| P04958 | |
| TETX | |
| E. coli-derived recombinant C. tetani TeNT Light Chain Pro2-Gly430 Accession # P04958 | |
| 100 μg | |
| Primary | |
| Detects TeNT Light Chain in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant TeNT Heavy Chain is observed. | |
| C. tetani | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 604023 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TeNT-LC, Tentoxylysin, Tetanus toxin chain H, Tetanus toxin chain L, Tetanus toxin heavy chain, Tetanus toxin light chain, tetX | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1061100 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction