Learn More
c-Fos Antibody (CL6381) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-76492JF646
Product Code. 30298768
  • 748.00€ / 0.10mL
Description
c-Fos Monoclonal antibody specifically detects c-Fos in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
c-Fos
Monoclonal
Janelia Fluor 646
activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, C-FOS, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, FOS, Fos proto-oncogene, AP-1 trancription factor subunit, G0/G1 switch regulatory protein 7, G0S7, p55, proto-oncogene c-Fos
This antibody was developed using a synthetic peptide derived from P01100, with the exact immunogen sequence remaining proprietary.
0.1 mL
Apoptosis, Cancer, Cellular Markers, Hypoxia, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Tumor Suppressors
2353
Store at 4⪚C in the dark.
IgG1
Western Blot
CL6381
50mM Sodium Borate
Mouse
Protein A purified