missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Fos Antibody (CL6381) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76492CL1
This item is not returnable.
View return policy
Description
c-Fos Monoclonal antibody specifically detects c-Fos in Human samples. It is validated for Western Blot
Specifications
| c-Fos | |
| Monoclonal | |
| CoraFluor 1 | |
| activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, C-FOS, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, FOS, Fos proto-oncogene, AP-1 trancription factor subunit, G0/G1 switch regulatory protein 7, G0S7, p55, proto-oncogene c-Fos | |
| This antibody was developed using a synthetic peptide derived from P01100, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cellular Markers, Hypoxia, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Tumor Suppressors | |
| 2353 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG1 |
| Western Blot | |
| CL6381 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction