missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
c-Fos Monoclonal antibody specifically detects c-Fos in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | c-Fos |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | CL6381 |
| Conjugate | Alexa Fluor 700 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, C-FOS, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, FOS, Fos proto-oncogene, AP-1 trancription factor subunit, G0/G1 switch regulatory protein 7, G0S7, p55, proto-oncogene c-Fos |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from P01100, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?