missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Fos Antibody (2G2), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
230.00€ - 568.00€
Specifications
| Antigen | c-Fos |
|---|---|
| Clone | 2G2 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18715283
|
Novus Biologicals
NBP2-37492 |
0.1 mL |
568.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633626
|
Novus Biologicals
NBP2-37492-0.025ml |
0.025 mL |
230.00€
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
c-Fos Monoclonal specifically detects c-Fos in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| c-Fos | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| activator protein 1, AP-1, cellular oncogene c-fos, Cellular oncogene fos, c-fos, FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS), FBJ murine osteosarcoma viral oncogene homolog, G0/G1 switch regulatory protein 7, G0S7, proto-oncogene c-Fos, v-fos FBJ murine osteosarcoma viral oncogene homolog | |
| FOS | |
| IgG1 | |
| Unpurified |
| 2G2 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cancer, Cellular Markers, Hypoxia, Neuronal Cell Markers, Neuroscience, Transcription Factors and Regulators, Tumor Suppressors | |
| Ascites with 0.03% Sodium Azide | |
| 2353 | |
| Purified recombinant fragment of human FOS expressed in E. Coli. | |
| Primary | |
| 40.7 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title