missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C/EBP alpha Monoclonal Antibody (692716)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561762
This item is not returnable.
View return policy
Description
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
The C/EBP (CCAAT/enhancer binding protein) family of transcription factors--alpha, beta, delta, and epsilon)--are widely expressed proteins found in a variety of tissues. These domains contain an evolutionarily conserved five-amino acid motif (the regulatory domain motif RDM) that conforms to the consensus sequence (I/V/L) KXEP. One of the major roles of these transcription factors is that of metabolic regulation in liver and fat cells. Studies suggest that these transcription factors act in a cascade aiding in the development of the adipose cell. Three C/EBP beta isoforms (LAP*, LAP, and LIP) arise from different translation initiation and display different function in gene regulation. From a neurological standpoint, C/EBPs have been implicated in the mediation of nerve growth factor, altering synaptic plasticity, and mediation of inflammatory response in AD patients.
Specifications
| C/EBP alpha | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P49715 | |
| CEBPA | |
| E. coli-derived recombinant human CEBP alpha , Met1-Ala124. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot, Immunocytochemistry | |
| 692716 | |
| Unconjugated | |
| CEBPA | |
| C EBP; C/EBP; c/EBP alpha; C/EBPa; C/ebpalpha; C/EBP-alpha; CAAT/enhancer-binding protein DNA-binding protein; CAAT/enhancer-binding protein, DNA-binding protein; CBF-A; CCAAT enhancer binding protein alpha; CCAAT/enhancer binding protein (C/EBP), alpha; CCAAT/enhancer binding protein alpha; CCAAT/enhancer binding protein, alpha; CCAAT/enhancer-binding protein alpha; Cebp; Cebpa; DBPCEP; OTTHUMP00000220269 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 1050 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction