missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Nitrosoadrenalone, Mikromol
Click to view available options
Quantity:
25 mg
Description
N-Nitrosoadrenalone, Mikromol
Specifications
Specifications
| Chemical Name or Material | N-Nitrosoadrenalone |
| Analyte or Component Names | N-Nitrosoadrenalone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H10 N2 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-[2-(3,4-dihydroxyphenyl)-2-oxo-ethyl]-N-methyl-nitrous amide, N-Nitrosoadrenalone |
| InChI Formula | InChI=1S/C9H10N2O4/c1-11(10-15)5-9(14)6-2-3-7(12)8(13)4-6/h2-4,12-13H,5H2,1H3 |
| Solution Type | Neat |
| SMILES | CN(CC(=O)c1ccc(O)c(O)c1)N=O |
| IUPAC Name | N-[2-(3,4-dihydroxyphenyl)-2-oxoethyl]-N-methylnitrous amide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction