Learn More
Buffer solution, Phthalate Potassium Hydrogen, pH 4.008, Fisher chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
1000 mL
Unit Size:
1000mL
500mL
Specifications
Specifications
| Chemical Name or Material | Buffer Solution, Potassium Hydrogen Phthalate |
| pH | 4.008 |
| Quantity | 1000 mL |
| Packaging | Bottle |