missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution pH 9, Chem Lab™
Click to view available options
Quantity:
1 L
Unit Size:
10 L
Specifications
Specifications
| Melting Point | 0°C |
| Color | Colorless |
| pH | 9 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Solution Type | Buffer |
| Formulation | H3BO3-KCl-NaOH |
| Odor | Odorless |
| Solubility Information | Water Soluble |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction