missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 8.0 (20°C), Buffer solution pH 8.0 (20°C), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
6 x 1 L
Unit Size:
Pack of 6
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Chemical Name or Material | Buffer solution pH 8.0 (20°C) |
| Name Note | Buffer solution pH 8.0 (20 °C) |
| Boiling Point | 100°C |
| CAS Min % | ≥90.0000%, ≥0.1000% |
| CAS Max % | ≤100.0000%, <1.0000% |
| CAS | 1330-43-4, 7647-01-0 |
| Density | 1.00 g/cm3 |
| Quantity | 6 x 1 L |
| Solution Type | Buffer |
| Linear Formula | C6H6 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction