Learn More
Color Coded pH Buffer Solution Tested at 25°C, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
1 L
5 L
Packaging:
Bag-in-Box Container
HDPE Bottle
HDPE Bottle GL45
Color:
Blue
Yellow
Unit Size:
1 L
5 L
500mL
Description
- Colored pH Buffer Solutions
- Tested at 25°C
- Certified by Reagecon’s ISO/IEC 17025 AccreditedTest Method
- NIST traceable
- Presented in various pack sizes
Specifications
Specifications
| Color | Blue |
| pH | 10 |
| Physical Form | Liquid |
| Quantity | 5 L |
| Packaging | HDPE Bottle |
Safety and Handling
Signal Word
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.