Learn More
Buffer solution pH 1.0 (20°C), Hydrochloric acid / potassium chloride, traceable to SRM from NIST and PTB, Honeywell Fluka™
Product Code. 15623180 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
6 x 1 L
Unit Size:
Pack of 6
Product Code. 15623180
Brand: Honeywell-Fluka 310446X1L
Request Bulk or Custom Format
Hydrochloric acid / potassium chloride, traceable to SRM from NIST and PTB
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Buffer solution pH 1.0 (20°C)
Name Note Hydrochloric acid / potassium chloride, traceable to SRM from NIST and PTB
Boiling Point 100°C
CAS Min % ≥99.0000%
CAS 7647-01-0, 7447-40-7
CAS Max % ≤0.5000%
Density 1.000 g/cm3
Quantity 6 x 1 L
Solution Type Buffer
Linear Formula CH3CH2OH
MDL Number MFCD00003568
Packaging Poly Bottle
UN Number UN1789
Synonym Hydrochloric acid – potassium chloride buffer solution
Show More Show Less
compliance-icons
Product Identifier
  • Buffer solution pH 1.0 (20 °C)
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P406-Store in corrosive resistant/ container with a resistant inner liner.
Supplemental information
  • MIXTURE LIST-Contains : Potassium chloride