Learn More
Buffer solution, Li2CO3 for AAS 0.02mg/L HNO3 matrix, Fisher chemical™
BIOPHARMA
Product Code. p-7083840 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 12970794
Brand: Fisher Chemical T 004 100
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Concentration 0.02 mg/L
Solvent or Matrix Nitric Acid
Quantity 100 mL
Chemical Name or Material Buffer solution, Li2CO3 for AAS
compliance-icons
Product Identifier
  • Ionisation buffer solution for AAS and ICP. Lithium (Li) 2% in HNO3 1% (T 004 )
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.