missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Sachets (NIST traceable), pH 10.01, Thermo Eutech™
Click to view available options
Quantity:
20 Ea.
Unit Size:
Pack of 20
Description
pH 10.01 buffer sachets (NIST traceable), box of 20 x 20 mL sachets
Economical NIST-traceable sachets for quick, accurate and convenient calibrations.
Simply insert electrode into sachet pack, calibrate, rinse and discard sachet – no separate container needed!
Specifications
Specifications
| Color | Colorless |
| pH | 10.01 |
| Quantity | 20 Ea. |
| Packaging | 20 mL sachet |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction