missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bub3 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
168.00€ - 507.00€
Specifications
| Antigen | Bub3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 10-20 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18584074
|
Novus Biologicals
NBP1-76796 |
0.1 mg |
507.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18585483
|
Novus Biologicals
NBP1-76796-0.025mg |
0.025 mg |
168.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Bub3 Polyclonal antibody specifically detects Bub3 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Bub3 | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 10-20 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| O43684 | |
| 9184 | |
| IgG | |
| Peptide affinity purified |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| PBS | |
| BUB3 (budding uninhibited by benzimidazoles 3, yeast) homolog, BUB3 budding uninhibited by benzimidazoles 3 homolog, BUB3L, budding uninhibited by benomyl, budding uninhibited by benzimidazoles 3 homolog (yeast), hBUB3, mitotic checkpoint component, mitotic checkpoint protein BUB3 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human bub3. The immunogen is located within the last 50 amino acids of Bub3. Amino Acid Squence: SYMYEMDDTEHPEDG | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title