Learn More
BTNL2/Butyrophilin-like 2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92488-0.1ml
This item is not returnable.
View return policy
Description
BTNL2/Butyrophilin-like 2 Polyclonal antibody specifically detects BTNL2/Butyrophilin-like 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| BTNL2/Butyrophilin-like 2 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:100 - 1:500, Immunohistochemistry-Paraffin 1:100 - 1:500 | |
| butyrophilin-like 2 (MHC class II associated) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 246-455 of human BTNL2 (NP_062548.1). LKVNGPSQPILVRVGEDIQLTCYLSPKANAQSMEVRWDRSHRYPAVHVYMDGDHVAGEQMAEYRGRTVLVSDAIDEGRLTLQILSARPSDDGQYRCLFEKDDVYQEASLDLKVVSLGSSPLITVEGQEDGEMQPMCSSDGWFPQPHVPWRDMEGKTIPSSSQALTQGSHGLFHVQTLLRVTNISAVDVTCSISIPFLGEEKIATFSLSGW | |
| 0.1 mL | |
| Immunology | |
| 56244 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |