missing translation for 'onlineSavingsMsg'
Learn More

BTN2A1 Antibody - Azide and BSA Free, Novus Biologicals™

Product Code. 18637511 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.02mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18637511 0.02 mL 0.02mL
18663201 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18637511 Supplier Novus Biologicals Supplier No. NBP2920200.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BTN2A1 Polyclonal antibody specifically detects BTN2A1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTN2A1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:1000
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias BK14H9.1, BTF1BT2.1butyrophilin BTF1, butyrophilin subfamily 2 member A1, butyrophilin, subfamily 2, member A1, DJ3E1.1, FLJ36567
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-240 of human BTN2A1 (NP_510961.1). FIVVGPTDPILATVGENTTLRCHLSPEKNAEDMEVRWFRSQFSPAVFVYKGGRERTEEQMEEYRGRTTFVSKDISRGSVALVIHNITAQENGTYRCYFQEGRSYDEAILHLVVAGLGSKPLISMRGHEDGGIRLECISRGWYPKPLTVWRDPYGGVAPALKEVSMPDADGLFMVTTAVIIRDKSVRNMSCSINNTLLGQKKESVIFIPESF
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11120
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.