missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTG2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97919-50ul
This item is not returnable.
View return policy
Description
BTG2 Polyclonal antibody specifically detects BTG2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| BTG2 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| B-cell translocation gene 2, BTG family member 2, BTG family, member 2, MGC126063, MGC126064, NGF-inducible proliferative protein PC3, PC3nerve growth factor-inducible proliferative, pheochromacytoma cell-3, protein BTG2, TIS21 | |
| Produced in rabbits immunized with E. coli-derived Human BTG2 fragment. | |
| 50 μg | |
| Cell Cycle and Replication | |
| 7832 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction