missing translation for 'onlineSavingsMsg'
Learn More

BRP44 Antibody (JCM-1), DyLight 755, Novus Biologicals™

Product Code. 18289116
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18289116 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18289116 Supplier Bio-Techne Supplier No. NBP280086IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BRP44 Monoclonal antibody specifically detects BRP44 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRP44
Applications Western Blot
Classification Monoclonal
Clone JCM-1
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias brain protein 44, DKFZP564B167, MGC125752, MGC125753
Host Species Mouse
Immunogen BRP44 peptide coupled to KLH.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25874
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.