Learn More
Brompheniramine Maleate, Mikromol™
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
Brompheniramine Maleate
Specifications
Specifications
| Chemical Name or Material | Brompheniramine maleate |
| CAS | 980-71-2 |
| Analyte or Component Names | Brompheniramine maleate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H19 Br N2 . C4 H4 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Brompheniramine Maleate |
| InChI Formula | InChI=1S/C16H19BrN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1- |
| SMILES | CN(C)CCC(c1ccc(Br)cc1)c2ccccn2.OC(=O)\C=C/C(=O)O |
| IUPAC Name | 3-(4-bromophenyl)-N,N-dimethyl-3-pyridin-2-ylpropan-1-amine;(Z)-but-2-enedioic acid |
| Show More |