missing translation for 'onlineSavingsMsg'
Learn More

Bromocriptine Mesilate, Mikromol™

Product Code. 17972686
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 17972686

Brand: Mikromol MM1159.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Bromocriptine Mesilate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromocriptine mesylate
CAS 22260-51-1
Analyte or Component Names Bromocriptine mesylate
Shipping Condition Room Temperature
Molecular Formula C32 H40 Br N5 O5 . C H4 O3 S
Certifications/Compliance ISO 17034
Synonym Bromocriptine mesilate, (6aR,9R)-5-Bromo-N-[(2R,5S,10aS,10bS)-10b-hydroxy-2-(1-methylethyl)-5-(2-methylpropyl)-3,6-dioxooctahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl]-7-methyl-4,6,6a,7,8,9-hexahydroindolo[4,3-fg]quinoline-9-carboxamide monomethanesulfonate, Ergocryptine, 2-bromo-, monomethanesulfonate (salt) (8CI), 8H-Oxazolo[3,2-a]pyrrolo[2,1-c]pyrazine, ergotaman-3',6',18-trione deriv., Indolo[4,3-fg]quinoline, ergotaman-3',6',18-trione deriv., (+)-Bromocriptine methanesulfonate, Bagren, Bromocriptine mesylate, Bromocriptine methanesulfonate, Bromocryptine mesylate, Bromocryptine methanesulfonate, CB 154, Cycloset, Parlodel, Pravidel, Ergotaman-3',6',18-trione, 2-bromo-12'-hydroxy-2'-(1-methylethyl)-5'-(2-methylpropyl)-, (5alpha)-, methanesulfonate (1:1), Ergotaman-3',6',18-trione, 2-bromo-12'-hydroxy-2'-(1-methylethyl)-5'-(2-methylpropyl)-, (5'alpha)-, monomethanesulfonate (salt) (9CI)
InChI Formula InChI=1S/C32H40BrN5O5.CH4O3S/c1-16(2)12-24-29(40)37-11-7-10-25(37)32(42)38(24)30(41)31(43-32,17(3)4)35-28(39)18-13-20-19-8-6-9-22-26(19)21(27(33)34-22)14-23(20)36(5)15-18;1-5(2,3)4/h6,8-9,13,16-18,23-25,34,42H,7,10-12,14-15H2,1-5H3,(H,35,39);1H3,(H,2,3,4)/t18-,23-,24+,25+,31-,32+;/m1./s1
SMILES CC(C)C[C@@H]1N2C(=O)[C@](NC(=O)[C@H]3CN(C)[C@@H]4Cc5c(Br)[nH]c6cccc(C4=C3)c56)(O[C@@]2(O)[C@@H]7CCCN7C1=O)C(C)C.CS(=O)(=O)O
IUPAC Name (6aR,9R)-5-Bromo-N-[(2R,5S,10aS,10bS)-10b-hydroxy-2-(1-methylethyl)-5-(2-methylpropyl)-3,6-dioxooctahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl]-7-methyl-4,6,6a,7,8,9-hexahydroindolo[4,3-fg]quinoline-9-carboxamide methanesulfonate
Molecular Weight (g/mol) 750.7
Quantity 100 mg
Formula Weight 749.2094 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.