missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocriptine Mesilate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Bromocriptine mesylate |
| CAS | 22260-51-1 |
| Analyte or Component Names | Bromocriptine mesylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C32 H40 Br N5 O5 . C H4 O3 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Bromocriptine mesilate, (6aR,9R)-5-Bromo-N-[(2R,5S,10aS,10bS)-10b-hydroxy-2-(1-methylethyl)-5-(2-methylpropyl)-3,6-dioxooctahydro-8H-oxazolo[3,2-a]pyrrolo[2,1-c]pyrazin-2-yl]-7-methyl-4,6,6a,7,8,9-hexahydroindolo[4,3-fg]quinoline-9-carboxamide monomethanesulfonate, Ergocryptine, 2-bromo-, monomethanesulfonate (salt) (8CI), 8H-Oxazolo[3,2-a]pyrrolo[2,1-c]pyrazine, ergotaman-3',6',18-trione deriv., Indolo[4,3-fg]quinoline, ergotaman-3',6',18-trione deriv., (+)-Bromocriptine methanesulfonate, Bagren, Bromocriptine mesylate, Bromocriptine methanesulfonate, Bromocryptine mesylate, Bromocryptine methanesulfonate, CB 154, Cycloset, Parlodel, Pravidel, Ergotaman-3',6',18-trione, 2-bromo-12'-hydroxy-2'-(1-methylethyl)-5'-(2-methylpropyl)-, (5alpha)-, methanesulfonate (1:1), Ergotaman-3',6',18-trione, 2-bromo-12'-hydroxy-2'-(1-methylethyl)-5'-(2-methylpropyl)-, (5'alpha)-, monomethanesulfonate (salt) (9CI) |
| InChI Formula | InChI=1S/C32H40BrN5O5.CH4O3S/c1-16(2)12-24-29(40)37-11-7-10-25(37)32(42)38(24)30(41)31(43-32,17(3)4)35-28(39)18-13-20-19-8-6-9-22-26(19)21(27(33)34-22)14-23(20)36(5)15-18;1-5(2,3)4/h6,8-9,13,16-18,23-25,34,42H,7,10-12,14-15H2,1-5H3,(H,35,39);1H3,(H,2,3,4)/t18-,23-,24+,25+,31-,32+;/m1./s1 |
| Solution Type | Neat |
| SMILES | CC(C)C[C@@H]1N2C(=O)[C@](NC(=O)[C@H]3CN(C)[C@@H]4Cc5c(Br)[nH]c6cccc(C4=C3)c56)(O[C@@]2(O)[C@@H]7CCCN7C1=O)C(C)C.CS(=O)(=O)O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction