missing translation for 'onlineSavingsMsg'
Learn More

BRN3B/POU4F2 Antibody, Novus Biologicals™

Product Code. 18708363 Shop All Bio Techne Products
Change view
Click to view available options
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
18708363 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18708363 Supplier Novus Biologicals Supplier No. NBP226061

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

BRN3B/POU4F2 Polyclonal specifically detects BRN3B/POU4F2 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Peptide ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRN3B/POU4F2
Applications Western Blot
Classification Polyclonal
Gene Symbols POU4F2
Purification Method Affinity Purified
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.