missing translation for 'onlineSavingsMsg'
Learn More

BRMS1 Antibody, Novus Biologicals™

Rabbit Monoclonal Antibody

213.00€ - 507.00€

Specifications

Antigen BRMS1
Dilution Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30233031
View Documents
Novus Biologicals
NBP3-33186-20ul
20 μL
213.00€
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30228773
View Documents
Novus Biologicals
NBP3-33186-100ul
100 μL
507.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BRMS1 Monoclonal antibody specifically detects BRMS1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications

Specifications

BRMS1
ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Cycle and Replication
PBS (pH 7.3), 50% glycerol, 0.05% BSA
25855
IgG
Affinity purified
Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Monoclonal
Purified
RUO
Human, Mouse, Rat
breast cancer metastasis suppressor 1, breast cancer metastasis-suppressor 1, DKFZP564A063
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human BRMS1 (Q9HCU9).,, Sequence:, MPVQPPSKDTEEMEAEGDSAAEMNGEEEESEEERSGSQTESEEESSEMDDEDYERRRSECVSEMLDLEKQFSELKEKLFRERLSQLRLRLEEVGAERAPE
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.