missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Breast cancer suppressor candidate 1 Antibody (OTI3D6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-02898
This item is not returnable.
View return policy
Description
Breast cancer suppressor candidate 1 Monoclonal specifically detects Breast cancer suppressor candidate 1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Breast cancer suppressor candidate 1 | |
| Monoclonal | |
| 0.69 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
| VWA5A | |
| Full length human recombinant protein of human VWA5A (NP_055437) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 4013 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI3D6 | |
| Unconjugated | |
| BCSC-1Loss of heterozygosity 11 chromosomal region 2 gene A protein, BCSC1ortholog of mouse AW551984, Breast cancer suppressor candidate 1, LOH11CR2Avon Willebrand factor A domain-containing protein 5A, loss of heterozygosity, 11, chromosomal region 2, gene A, von Willebrand factor A domain containing 5A | |
| Mouse | |
| 86.3 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction