missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BrdU Monoclonal Antibody (A9C7)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550620
This item is not returnable.
View return policy
Description
Positive control: BrdU treated mouse embryo tissue, BrdU treated Hela, BrdU treated NIH/3T3, EdU treated mouse embryo tissue, EdU treated Hela, EdU treated NIH/3T3. Subcellular Location: Nucleus.
Bromodeoxyuridine (5-bromo-2-deoxyuridine, BrdU) is a synthetic nucleoside that is an analogue of thymidine. BrdU is commonly used in the detection of proliferating cells in living tissues, and can be incorporated into the newly synthesized DNA of replicating cells (during the S phase of the cell cycle), substituting for thymidine during DNA replication. Antibodies specific for BrdU can then be used to detect the incorporated chemical, thus indicating cells that were actively replicating their DNA. Binding of the antibody requires denaturation of the DNA, usually by exposing the cells to acid or heat.
Specifications
| BrdU | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical, Mouse | |
| Antibody | |
| IgG1 |
| Dot Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| A9C7 | |
| Unconjugated | |
| 5-Bromo-2-deoxyuridine; Bromodeoxyuridine | |
| BrdU-OVA. | |
| 100 μL | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction