missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Bovine Serum Albumin Native Protein
Shop All Bio Techne Products
Click to view available options
:
1mg; Texas Red
Description
Native Bovine Serum Albumin. Source: Bovine Serum The Bovine Serum Albumin Native Protein is derived from Bovine Serum. The Bovine Serum Albumin Native Protein has been validated for the following applications: Western Blot, Dot Blot, Fluorophore-linked immunosorbent assay, Immunofluorescence.
Specifications
Specifications
| For Use With (Application) | Dot Blot |
| Formulation | Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10mg/mL Polyethylene Glycol (PEG-8000) |
| Gene ID (Entrez) | 280717 |
| Name | Bovine Serum Albumin protein |
| Storage Requirements | 4°C in the dark. |
| Cross Reactivity | Bovine |
| Purity or Quality Grade | Multi-step |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction