Learn More
Bosentan USP Related Compound D, Mikromol™
Description
Bosentan USP Related Compound D
Specifications
Specifications
| Chemical Name or Material | 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine |
| CAS | 150728-13-5 |
| Analyte or Component Names | 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H10 Cl2 N4 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2,2'-Bipyrimidine, 4,6-dichloro-5-(2-methoxyphenoxy)-, 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine, 4,6-Dichloro-5-(2-methoxyphenoxy)-2,2'-dipyrimidine, 4,6-Dichloro-5-(o-methoxyphenoxy)-2-(2-pyrimidinyl)pyrimidine, (4,6-dichloro-5-(2-methoxyphenoxy)-2,2'-bipyrimidine), Bosentan Related Compound D, Bosentan Related Compound D |
| InChI Formula | InChI=1S/C15H10Cl2N4O2/c1-22-9-5-2-3-6-10(9)23-11-12(16)20-15(21-13(11)17)14-18-7-4-8-19-14/h2-8H,1H3 |
| SMILES | COc1ccccc1Oc2c(Cl)nc(nc2Cl)c3ncccn3 |
| IUPAC Name | 4,6-dichloro-5-(2-methoxyphenoxy)-2-pyrimidin-2-ylpyrimidine |
| Show More |
Product Title
Haben Sie Verbesserungsvorschläge?