Learn More
Boron, AAS solution standard, Specpure™ B 1000μg/mL
Product Code. 11316339
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 11316339
Brand: Thermo Scientific Alfa Aesar 088055.AP
Request Bulk or Custom Format
H3BO3 in H2O
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
Molecular Formula H3BO3 in H2O
MDL Number MFCD00134034
Specifications
Physical Form Liquid
Quantity 500 mL
Grade Specpure
Chemical Name or Material Boron, AAS standard solution, B 1000μg/mL