missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BNP Antibody (8D5B4C11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47507-0.025ml
This item is not returnable.
View return policy
Description
BNP Monoclonal specifically detects BNP in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| BNP | |
| Monoclonal | |
| Unconjugated | |
| BNP, brain type natriuretic peptide, Gamma-brain natriuretic peptide, natriuretic peptide B, natriuretic peptide precursor B, natriuretic peptides B, natriuretic protein | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
| ELISA | |
| 8D5B4C11 | |
| ELISA 1:10000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:1000 | |
| NPPB | |
| Synthetic peptide corresponding to aa (Cys-Phe-Gly-Arg-Lys-Met-Asp-Arg-Ile-Ser-Ser-Ser-Ser) of human BNP, conjugated to KLH. | |
| 0.025 mL | |
| Cytokine Research, Immunology, Neuroscience, Neurotransmission | |
| 4879 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction