missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BNIP1 Monoclonal specifically detects BNIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
| Antigen | BNIP1 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI2B3 |
| Conjugate | Biotin |
| Dilution | Western Blot, Immunohistochemistry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | BCL2/adenovirus E1B 19 kDa protein-interacting protein 1, BCL2/adenovirus E1B 19kDa interacting protein 1, BCL2/adenovirus E1B 19kD-interacting protein 1, NIP1, SEC20, SEC20L, Transformation-related gene 8 protein, TRG-8, vesicle transport protein SEC20 |
| Gene Symbols | BNIP1 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?