Learn More
BNA Internal Standard Mixture 990 2000 μg/mL in Methanol, Dr. Ehrenstorfer
Product Code. 30319018
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319018 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30319018 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000990ME
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
CAS 3017-95-6, 462-06-6
Quantity 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol