missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BMP1 Monoclonal Antibody (264822)
GREENER_CHOICE

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561067

Product Code. 30534754

  • 589.00€ / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

This gene encodes a protein that is capable of inducing formation of cartilage in vivo. Although other bone morphogenetic proteins are members of the TGF-beta superfamily, this gene encodes a protein that is not closely related to other known growth factors. This gene is expressed as alternatively spliced variants that share an N-terminal protease domain but differ in their C-terminal region.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BMP1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P13497
BMP1
Mouse myeloma cell line NS0-derived recombinant human BMP-1/PCP, Ala121-Gln730.
500 μg
Primary
Human
Antibody
IgG2b
Immunoprecipitation, Western Blot
264822
Unconjugated
BMP1
BMP; Bmp1; BMP-1; Bone morphogenetic protein; Bone morphogenetic protein 1; FLJ44432; mammalian tolloid protein; mTld; OI13; PCOLC; Pcp; PCP2; pCP-2; procollagen C-endopeptidase; procollagen C-proteinase; procollagen C-proteinase 3; TLD
Rat
Protein A/G
RUO
649
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.