missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bmf Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
178.00€
Specifications
| Antigen | Bmf |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunofluorescence 5 μg/mL, Immunocytochemistry 10 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Bmf Polyclonal specifically detects Bmf in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Bmf | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunofluorescence 5 μg/mL, Immunocytochemistry 10 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Tumor Suppressors | |
| PBS with 0.02% Sodium Azide | |
| Bcl2 modifying factor, bcl-2-modifying factor, FLJ00065 | |
| BMF | |
| IgG | |
| Affinity Purified | |
| 22 kDa |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| NP_277038 | |
| 90427 | |
| Antibody was raised with a synthetic peptide corresponding to 15 amino acids near the amino terminus of human Bmf. The immunogen is located within the first 50 amino acids of Bmf. Amino Acid Squence: EPSQCVEELEDDV | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title