Learn More
3M™ Blue Disposable Visitor Lab Coats, Series 4400
Product Code. p-8003408
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 50
Product Code. 12991325
Brand: 3M™ 7000089701
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Use for hygiene applications. The 3M Disposable Visitor Lab Coats series is made from lightweight polypropylene.
  • Ideal for hygiene applications
  • 100% soft, breathable, and lightweight polypropylene
  • Zip fastener
  • Internal pocket
TRUSTED_SUSTAINABILITY
Specifications
Product Type Lab Coat
Size Large
Material Polypropylene
Color Blue
Closure Type Zipper
Cuff Style Elastic
Disposable Single-use
Seam Type Serged
Size (English) Chest 36 to 38 in.
Size (Metric) Chest 91 to 97 cm
Certifications/Compliance CE Category 1
For Use With (Application) Non-hazardous liquids and dust
Includes Internal pocket on the wearers left hand side (measuring 13 H x 12cm W)
Packaging Type Individual packaging