Learn More
Witeg™ Blue Borosilicate Glass Laboratory Bottles
Product Code. 11827860
Click to view available options
Capacity (Metric):
100 mL
1000 mL
2000 mL
Unit Size:
Pack of 10
Product Code. 11827860
Brand: Witeg™ 5 526 100 BL
Only null left
Add to basket
Add to basket
Store, mix and blend your media with Witeg™ Blue Borosilicate Laboratory Bottles, which come in various capacities.
  • Blue graduation markings
  • Borosilicate glass
Specifications
Capacity (Metric) 100 mL
Color Clear
For Use With (Application) Laboratory
Material Polyethylene
Closure Color Blue
Graduations Yes
Certifications/Compliance ISO 4796
Closure Included Yes
Mouth Wide
Product Type Laboratory Bottle
Closure Material Polypropylene
Closure Type Screw Cap