missing translation for 'onlineSavingsMsg'
Learn More

BLOC1S2 Antibody (8C10), Novus Biologicals™

Product Code. 18531062 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18531062 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18531062 Supplier Novus Biologicals Supplier No. H00282991M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BLOC1S2 Monoclonal antibody specifically detects BLOC1S2 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen BLOC1S2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 8C10
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_776170.2
Gene Alias biogenesis of lysosomal organelles complex-1, subunit 2, BLOC-1 subunit 2, CEAP, centrosomal 10 kDa protein, centrosome protein oncogene, Centrosome-associated protein, subunit 2
Host Species Mouse
Immunogen BLOC1S2 (NP_776170.2, 1 a.a. ∽ 142 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MAAAAEGVLATRSDEPARDDAAVETAEEAKEPAEADITELCRDMFSKMATYLTGELTATSEDYKLLENMNKLTSLKYLEMKDIAINISRNLKDLNQKYAGLQPYLDQINVIEEQVAALEQAAYKLDAYSKKLEAKYKKLEKR
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 282991
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.