Learn More
SCAT Blind Plugs for HPLC
Product Code. p-7225782 Shop All SCAT Europe Products
Change view
Click to view available options
For Use With:
Exhaust Filter
Ground Neck Bottles w/ Locknut
Safety Caps, Safety Waste Caps
Color:
Colorless
White
Material:
PFA
PTFE
Thread Size:
GL14
NPT 1/8 in.
NS29/32
UNF 1/4 in. 28G
Unit Size:
Each
Pack of 10
Product Code. For Use With Color Material Thread Size unitSize
10226504 Exhaust Filter White PTFE GL14 Each
13233859 Safety Caps, Safety Waste Caps Colorless PFA UNF 1/4 in. 28G Pack of 10
10051453 Safety Caps, Safety Waste Caps Colorless PTFE NPT 1/8 in. Each
10357579 Ground Neck Bottles w/ Locknut White PTFE NS29/32 Each
4 options
Product Code. 10226504
Brand: SCAT 107620
Blind plugs for use with HPLC instruments.
Be prepared for HPLC with these SCAT™ blind plugs.
Specifications
Type Blind Plug
Color White
For Use With (Application) HPLC/UHPLC Systems
Thread Size GL14
For Use With Exhaust Filter
Material PTFE