missing translation for 'onlineSavingsMsg'
Learn More

Bisphenol A diallyl ether, Dr. Ehrenstorfer

Product Code. 30327906
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327906 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327906 Supplier Dr. Ehrenstorfer Supplier No. DREC10655615

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bisphenol A Diallyl Ether
CAS 3739-67-1
Analyte or Component Names Bisphenol A Diallyl Ether
Shipping Condition +20°C
Molecular Formula C21 H24 O2
Certifications/Compliance ISO/IEC 17025
Synonym 1,1'-(1-Methylethylidene)bis[4-(2-propen-1-yloxy)benzene] (ACI), Benzene, 1,1'-(1-methylethylidene)bis[4-(2-propenyloxy)- (9CI), Propane, 2,2-bis[p-(allyloxy)phenyl]- (6CI, 7CI, 8CI), 1-Prop-2-enoxy-4-[2-(4-prop-2-enoxyphenyl)propan-2-yl]benzene, 2,2-Bis(4-allyloxyphenyl)propane, 4,4'-Isopropylidenebis[(allyloxy)benzene], 4,4'-Isopropylidenediphenol diallyl ether, 912A, B 303589
InChI Formula InChI=1S/C21H24O2/c1-5-15-22-19-11-7-17(8-12-19)21(3,4)18-9-13-20(14-10-18)23-16-6-2/h5-14H,1-2,15-16H2,3-4H3
SMILES CC(C)(c1ccc(OCC=C)cc1)c2ccc(OCC=C)cc2
IUPAC Name 1-prop-2-enoxy-4-[2-(4-prop-2-enoxyphenyl)propan-2-yl]benzene
Molecular Weight (g/mol) 308.4141
Quantity 100 mg
Formula Weight 308.1776 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.