Learn More
Bis(tert-butylcyclopentadienyl)hafnium(IV) dichloride, 98+%
Product Code. 11426600
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11426600
Brand: Thermo Scientific Alfa Aesar H27638.06
Request Bulk or Custom Format
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
CAS: 33010-55-8 | C18H26Cl2Hf | 491.796 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 33010-55-8
Molecular Formula C18H26Cl2Hf
Molecular Weight (g/mol) 491.796
MDL Number MFCD01073803
InChI Key ZZDLUYCFDDVDOX-UHFFFAOYSA-L
Synonym hafnium chloride 2-tert-butylcyclopenta-1,3-dien-1-ide 1/2/2
PubChem CID 74765427
IUPAC Name 2-tert-butylcyclopenta-1,3-diene;hafnium(4+);dichloride
SMILES CC(C)(C)C1=[C-]CC=C1.CC(C)(C)C1=[C-]CC=C1.[Cl-].[Cl-].[Hf+4]
Specifications
Melting Point 195°C
Quantity 5 g
Sensitivity Air and moisture sensitive
Formula Weight 491.8
Percent Purity ≥98%
Chemical Name or Material Bis(tert-butylcyclopentadienyl)hafnium(IV) dichloride