accessibility menu, dialog, popup

UserName

CAS RN 33010-55-8

CH3H3CCH3CCH3H3CCH3CClClHf

CAS RN 33010-55-8

IUPAC Name: hafnium(4+) bis(2-tert-butylcyclopenta-1,3-dien-1-ide) dichloride
Synonyms: hafnium(4+) bis(2-tert-butylcyclopenta-1,3-dien-1-ide) dichloride
Molecular Weight (g/mol): 491.8
Molecular Formula: C18H26Cl2Hf
InChi Key: ZZDLUYCFDDVDOX-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Hf+4].CC(C)(C)C1=[C-]CC=C1.CC(C)(C)C1=[C-]CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results