missing translation for 'onlineSavingsMsg'
Learn More

Bis(4-fluorophenyl) ether, 98%, Thermo Scientific™

Product Code. 11367885
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367885 5 g 5g
11357885 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367885 Supplier Thermo Scientific Alfa Aesar Supplier No. L02885.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 330-93-8
Molecular Formula C12H8F2O
Molecular Weight (g/mol) 206.19
MDL Number MFCD00013552
InChI Key UUKHFGSOCZLVJO-UHFFFAOYSA-N
Synonym bis 4-fluorophenyl ether, 4,4'-oxybis fluorobenzene, 4-fluorophenyl ether, 4,4'-difluorodiphenyl ether, 1-fluoro-4-4-fluorophenoxy benzene, 1,1'-oxybis 4-fluorobenzene, unii-9r9o553twz, p-fluorophenyl ether, benzene, 1,1'-oxybis 4-fluoro
PubChem CID 67615
IUPAC Name 1-fluoro-4-(4-fluorophenoxy)benzene
SMILES FC1=CC=C(OC2=CC=C(F)C=C2)C=C1

Specifications

Density 1.23
Boiling Point 240°C
Flash Point >110°C (230°F)
Odor Characteristic
Refractive Index 1.5375
Quantity 5 g
Beilstein 2559168
Formula Weight 206.19
Percent Purity 98%
Chemical Name or Material Bis(4-fluorophenyl) ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.