accessibility menu, dialog, popup

UserName

CAS RN 330-93-8

IUPAC Name: 1-fluoro-4-(4-fluorophenoxy)benzene
Synonyms: 1-fluoro-4-(4-fluorophenoxy)benzene
Molecular Weight (g/mol): 206.19
Molecular Formula: C12H8F2O
InChi Key: UUKHFGSOCZLVJO-UHFFFAOYSA-N
SMILES: FC1=CC=C(OC2=CC=C(F)C=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results