missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Birch pollen Bet V1, Rabbit anti-Plant, Clone: BV16, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81093-0.2MG
This item is not returnable.
View return policy
Description
Birch pollen Bet V1 Monoclonal specifically detects Birch pollen Bet V1 in Plant samples. It is validated for Western Blot, ELISA.
Specifications
| Birch pollen Bet V1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Allergen Bet v I-A, Allergen=Bet v 1-A, BETVI, BETVIA, Major pollen allergen Bet v 1-A | |
| Birch pollen Bet V1-A, a major allergen of the European white birch. | |
| 0.2 mg | |
| Primary | |
| Plant | |
| Purified |
| Western Blot, ELISA | |
| BV16 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| This antibody binds to Birch pollen Bet V1. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction