Learn More
Biphenamine Hydrochloride, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Biphenamine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Biphenamine Hydrochloride |
| CAS | 5560-62-3 |
| Analyte or Component Names | Biphenamine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H23 N O3 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | [1,1'-Biphenyl]-3-carboxylic acid, 2-hydroxy-, 2-(diethylamino)ethyl ester, hydrochloride (1:1), 3-Biphenylcarboxylic acid, 2-hydroxy-, 2-(diethylamino)ethyl ester, hydrochloride (7 CI,8 CI), [1,1'-Biphenyl]-3-carboxylic acid, 2-hydroxy-, 2-(diethylamino)ethyl ester, hydrochloride (9 CI), 2-(Diethylamino)ethyl 2-hydroxy-3-biphenylcarboxylate hydrochloride, 2-(Diethylamino)ethyl 3-Phenylsalicylate hydrochloride, Biphenamine hydrochloride, Melsaphine, Sebaclen, Sebaklen, Xenylsalate hydrochloride |
| InChI Formula | InChI=1 S/C19H23NO3.ClH/c1-3-20(4-2)13-14-23-19(22)17-12-8-11-16(18(17)21)15-9-6-5-7-10-15;/h5-12,21 H,3-4,13-14H2,1-2H3;1 H |
| SMILES | Cl.CCN(CC)CCOC(=O)c1cccc(c1O)c2ccccc2 |
| IUPAC Name | 2-(diethylamino)ethyl 2-hydroxy-3-phenylbenzoate;hydrochloride |
| Show More |